siRNA Details
Gene Symbol: Tom1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253859.1 | NP_001240788.1 | 8, 9 | 842 | ||
XM_008767967.1 | XP_008766189.1 | 7, 8 | 769 | |||
GenBank mRNA | BC169112.1 | - | 8, 9 | 840 |
