siRNA Details
Gene Symbol: Slc35b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199081.1 | NP_954512.1 | 7, 8 | 850 | ||
GenBank mRNA | AY349135.1 | - | 7, 8 | 850 | ||
BC085347.1 | - | 7, 8 | 885 | 7305181 | ||
D87991.1 | - | 7, 8 | 843 | |||
FQ233464.1 | - | 7, 8 | 873 |
