siRNA Details
Gene Symbol: Helz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105848.2 | NP_001099318.2 | 24 | 3642 | ||
XM_006247562.2 | XP_006247624.1 | 26 | 3763 | |||
XM_006247565.2 | XP_006247627.1 | 26 | 3760 | |||
XM_008768314.1 | XP_008766536.1 | 27 | 3814 | |||
XM_008768315.1 | XP_008766537.1 | 27 | 3811 | |||
XM_008768316.1 | XP_008766538.1 | 25 | 3706 |
