siRNA Details
Gene Symbol: Abca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768405.1 | XP_008766627.1 | 5 | 1536 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768405.1 | XP_008766627.1 | 5 | 1536 |