siRNA Details
Gene Symbol: Cd300lf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025111.1 | NP_001020282.1 | 7 | 906 | ||
XM_008768332.1 | XP_008766554.1 | 8 | 1819 | |||
XM_008768333.1 | XP_008766555.1 | 9 | 1348 | |||
XM_008768334.1 | XP_008766556.1 | 8 | 1238 | |||
XM_008768335.1 | XP_008766557.1 | 7 | 1301 | |||
XM_008768336.1 | XP_008766558.1 | 7 | 1067 | |||
XM_008768337.1 | XP_008766559.1 | 8 | 1798 | |||
XM_008768338.1 | XP_008766560.1 | 8 | 1751 | |||
XM_008768339.1 | XP_008766561.1 | 8 | 1729 | |||
XM_008768340.1 | XP_008766562.1 | 7 | 1693 | |||
XM_008768341.1 | XP_008766563.1 | 8 | 1661 | |||
XM_008768342.1 | XP_008766564.1 | 7 | 1601 | |||
GenBank mRNA | BC093593.1 | - | 7 | 906 | 7314621 | |
GU057984.1 | - | 7 | 781 | |||
GU062903.1 | - | 6 | 721 | |||
HQ263412.1 | - | 6 | 801 |
