siRNA Details
Gene Symbol: Cd300lf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025111.1 | NP_001020282.1 | 1 | 71 | ||
XM_008768332.1 | XP_008766554.1 | 2 | 927 | |||
XM_008768333.1 | XP_008766555.1 | 3 | 456 | |||
XM_008768334.1 | XP_008766556.1 | 2 | 346 | |||
XM_008768337.1 | XP_008766559.1 | 2 | 927 | |||
XM_008768338.1 | XP_008766560.1 | 2 | 925 | |||
XM_008768339.1 | XP_008766561.1 | 2 | 921 | |||
XM_008768340.1 | XP_008766562.1 | 2 | 924 | |||
XM_008768341.1 | XP_008766563.1 | 2 | 919 | |||
XM_008768342.1 | XP_008766564.1 | 2 | 916 | |||
GenBank mRNA | BC093593.1 | - | 1 | 71 | 7314621 | |
GU057984.1 | - | 1 | 39 | |||
GU062903.1 | - | 1 | 36 | |||
HQ263412.1 | - | 1 | 39 |
