siRNA Details
Gene Symbol: Cd300lf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025111.1 | NP_001020282.1 | 7 | 1005 | ||
XM_008768332.1 | XP_008766554.1 | 8 | 1918 | |||
XM_008768333.1 | XP_008766555.1 | 9 | 1447 | |||
XM_008768334.1 | XP_008766556.1 | 8 | 1337 | |||
XM_008768335.1 | XP_008766557.1 | 7 | 1400 | |||
XM_008768336.1 | XP_008766558.1 | 7 | 1166 | |||
XM_008768337.1 | XP_008766559.1 | 8 | 1897 | |||
XM_008768338.1 | XP_008766560.1 | 8 | 1850 | |||
XM_008768339.1 | XP_008766561.1 | 8 | 1828 | |||
XM_008768340.1 | XP_008766562.1 | 7 | 1792 | |||
XM_008768341.1 | XP_008766563.1 | 8 | 1760 | |||
XM_008768342.1 | XP_008766564.1 | 7 | 1700 | |||
GenBank mRNA | BC093593.1 | - | 7 | 1005 | 7314621 | |
GU057984.1 | - | 7 | 880 | |||
GU062903.1 | - | 6 | 820 | |||
HQ263412.1 | - | 6 | 900 |
