siRNA Details
Gene Symbol: Hid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247774.2 | XP_006247836.1 | 4, 5 | 632 | ||
XM_213523.8 | XP_213523.6 | 4, 5 | 628 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247774.2 | XP_006247836.1 | 4, 5 | 632 | ||
XM_213523.8 | XP_213523.6 | 4, 5 | 628 |