siRNA Details
Gene Symbol: Fbf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247750.2 | XP_006247812.1 | 4, 5 | 845 | ||
XM_006247759.2 | XP_006247821.1 | 4, 5 | 269 | |||
XM_006247760.2 | XP_006247822.1 | 4, 5 | 845 | |||
XM_006247761.2 | XP_006247823.1 | 4, 5 | 845 | |||
XM_008768413.1 | XP_008766635.1 | 4, 5 | 842 | |||
XM_008768414.1 | XP_008766636.1 | 4, 5 | 845 | |||
XM_008768415.1 | XP_008766637.1 | 4, 5 | 846 | |||
XM_008768416.1 | XP_008766638.1 | 5, 6 | 920 | |||
XM_008768417.1 | XP_008766639.1 | 5, 6 | 348 | |||
XM_008768418.1 | XP_008766640.1 | 246 | ||||
XM_008768419.1 | XP_008766641.1 | 5, 6 | 917 | |||
XM_008768420.1 | XP_008766642.1 | 4, 5 | 846 | |||
GenBank mRNA | FQ210392.1 | - | 4, 5 | 207 |
