siRNA Details
Gene Symbol: Ccdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134688.1 | NP_001128160.1 | 5 | 1567 | ||
XM_008768470.1 | XP_008766692.1 | 4 | 998 | |||
XM_008768471.1 | XP_008766693.1 | 4 | 995 |
