siRNA Details
Gene Symbol: Adamts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137622.1 | NP_001131094.1 | 16, 17 | 2526 | ||
XM_008767677.1 | XP_008765899.1 | 14, 15 | 2357 | |||
XM_008767678.1 | XP_008765900.1 | 14, 15 | 2357 |
