siRNA Details
Gene Symbol: Dgcr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105865.1 | NP_001099335.1 | 6 | 1835 | ||
XM_006248606.2 | XP_006248668.1 | 5 | 1662 | |||
XM_006248607.2 | XP_006248669.1 | 5 | 5361 | |||
XM_008768865.1 | XP_008767087.1 | 4 | 5764 |
