siRNA Details
Gene Symbol: Ccdc14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727893.3 | XP_002727939.1 | 13 | 2736 | ||
XM_006248441.2 | XP_006248503.1 | 9 | 3250 | |||
XM_008768770.1 | XP_008766992.1 | 9 | 3165 | |||
XM_008768771.1 | XP_008766993.1 | 9 | 2314 |
