siRNA Details
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008286.1 | NP_001008287.1 | 10 | 893 | ||
XM_006234370.2 | XP_006234432.1 | 10 | 724 | |||
GenBank mRNA | BC086360.1 | - | 10 | 893 | 7303884 | |
FQ211359.1 | - | 1 | 51 | |||
FQ213350.1 | - | 10 | 867 | |||
FQ215122.1 | - | 10 | 861 | |||
FQ215507.1 | - | 10 | 887 | |||
FQ215582.1 | - | 10 | 911 |
