siRNA Details
Gene Symbol: Mtx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008286.1 | NP_001008287.1 | 9 | 751 | ||
XM_006234370.2 | XP_006234432.1 | 9 | 582 | |||
XM_006234371.2 | XP_006234433.1 | 9 | 784 | |||
GenBank mRNA | BC086360.1 | - | 9 | 751 | 7303884 | |
FQ213350.1 | - | 9 | 725 | |||
FQ215122.1 | - | 9 | 719 | |||
FQ215582.1 | - | 9 | 769 |
