siRNA Details
Gene Symbol: Cct8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105897.2 | NP_001099367.2 | 4, 5 | 513 | ||
GenBank mRNA | CV076369.1 | - | 1, 2 | 91 | ||
DY312304.1 | - | 4, 5 | 513 | |||
FQ225223.1 | - | 4, 5 | 486 | |||
FQ226011.1 | - | 4, 5 | 486 |
