siRNA Details
Gene Symbol: Uspl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105906.2 | NP_001099376.1 | 4 | 867 | ||
NM_001198555.1 | NP_001185484.1 | 4 | 867 | |||
XM_006248810.2 | XP_006248872.1 | 3 | 810 | |||
GenBank mRNA | DY309833.1 | - | 1 | 288 |
