siRNA Details
Gene Symbol: Uspl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105906.2 | NP_001099376.1 | 3 | 317 | ||
NM_001198555.1 | NP_001185484.1 | 3 | 317 | |||
XM_006248810.2 | XP_006248872.1 | 2 | 260 | |||
GenBank mRNA | CB712845.1 | - | 2 | 148 | ||
CB798160.1 | - | 2 | 151 |
