siRNA Details
Gene Symbol: LOC103690050 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751116.3 | XP_003751164.1 | 7 | 836 | ||
XM_006248816.2 | XP_006248878.1 | 7 | 1122 | |||
XM_006248817.2 | XP_006248879.1 | 7 | 855 |
Gene Symbol: Katnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006956.1 | NP_001006957.1 | 836 | |||
XM_006248824.2 | XP_006248886.1 | 1122 | ||||
GenBank mRNA | BC083673.1 | - | 836 | 7135708 |
