siRNA Details
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105908.1 | NP_001099378.1 | 5 | 495 | ||
XM_006248862.2 | XP_006248924.1 | 5 | 490 | |||
GenBank mRNA | BC160858.1 | - | 5 | 429 | 8365723 |
