siRNA Details
Gene Symbol: Sbds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008289.1 | NP_001008290.1 | 1 | 112 | ||
GenBank mRNA | BC086335.1 | - | 1 | 112 | 7301062 |

More Information
Other Species Targeted

Gene Ontology Categories:
