siRNA Details
Gene Symbol: Rab35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013046.1 | NP_001013064.1 | 3, 4 | 324 | ||
XM_008769221.1 | XP_008767443.1 | 3, 4 | 374 | |||
GenBank mRNA | BC085769.1 | - | 3, 4 | 324 | 7135343 |
