siRNA Details
Gene Symbol: Chfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009258.1 | NP_001009258.1 | 11 | 1442 | ||
XM_006249535.2 | XP_006249597.1 | 11 | 1521 | |||
XM_006249536.2 | XP_006249598.1 | 11 | 1521 | |||
GenBank mRNA | BC087162.1 | - | 11 | 1442 | 7135077 |
