siRNA Details
Gene Symbol: Foxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105935.1 | NP_001099405.1 | 6 | 624 | ||
XM_006249493.2 | XP_006249555.1 | 7 | 1030 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Foxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105935.1 | NP_001099405.1 | 6 | 624 | ||
XM_006249493.2 | XP_006249555.1 | 7 | 1030 |