siRNA Details
Gene Symbol: Cc2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013869.1 | NP_001013891.1 | 24 | 2537 | ||
XM_006255233.2 | XP_006255295.1 | 25 | 2729 | |||
XM_006255234.2 | XP_006255296.1 | 25 | 2726 | |||
XM_008772364.1 | XP_008770586.1 | 24 | 2595 | |||
GenBank mRNA | BC081948.1 | - | 24 | 2537 | 7122187 |
