siRNA Details
Gene Symbol: Slc35f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105950.1 | NP_001099420.1 | 4 | 513 | ||
XM_006249671.2 | XP_006249733.1 | 4 | 410 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc35f5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105950.1 | NP_001099420.1 | 4 | 513 | ||
XM_006249671.2 | XP_006249733.1 | 4 | 410 |