siRNA Details
Gene Symbol: Lrrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177368.1 | NP_001170839.1 | 2 | 2015 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177368.1 | NP_001170839.1 | 2 | 2015 |