siRNA Details
Gene Symbol: Adipor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207587.1 | NP_997470.1 | 8 | 1329 | ||
XM_006249852.2 | XP_006249914.1 | 8 | 1365 | |||
XM_008769543.1 | XP_008767765.1 | 8 | 1666 | |||
XM_008769544.1 | XP_008767766.1 | 8 | 2028 | |||
GenBank mRNA | BC061838.1 | - | 8 | 1329 | 5599938 | |
FQ217287.1 | - | 1 | 111 | |||
FQ233463.1 | - | 1 | 101 |
