siRNA Details
Gene Symbol: Fmo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769727.1 | XP_008767949.1 | 2 | 158 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fmo13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769727.1 | XP_008767949.1 | 2 | 158 |