siRNA Details
Gene Symbol: Lrrc52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077434.1 | NP_001070902.1 | 1 | 821 | ||
GenBank mRNA | BC126099.1 | - | 1 | 821 | 7112275 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrc52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077434.1 | NP_001070902.1 | 1 | 821 | ||
GenBank mRNA | BC126099.1 | - | 1 | 821 | 7112275 |