siRNA Details
Gene Symbol: Slamf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105973.1 | NP_001099443.1 | 4 | 817 | ||
XM_006250284.2 | XP_006250346.1 | 4 | 2230 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slamf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105973.1 | NP_001099443.1 | 4 | 817 | ||
XM_006250284.2 | XP_006250346.1 | 4 | 2230 |