siRNA Details
Gene Symbol: Tmem63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134496.2 | NP_001127968.2 | 11 | 1019 | ||
XM_006250364.2 | XP_006250426.1 | 11 | 1196 | |||
XM_006250365.2 | XP_006250427.1 | 11 | 1051 | |||
GenBank mRNA | FQ210846.1 | - | 11 | 991 |
