siRNA Details
Gene Symbol: Tlr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145828.1 | NP_001139300.1 | 1 | 3227 | ||
XM_006250376.1 | XP_006250438.1 | 5 | 3499 | |||
XM_006250377.2 | XP_006250439.1 | 4 | 3752 | |||
XM_008769824.1 | XP_008768046.1 | 2 | 3091 | |||
XM_008769825.1 | XP_008768047.1 | 2 | 5454 | |||
XM_008769826.1 | XP_008768048.1 | 4 | 4822 | |||
GenBank mRNA | FJ750588.1 | - | 1 | 2260 |
