siRNA Details
Gene Symbol: Tlr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145828.1 | NP_001139300.1 | 1 | 1333 | ||
XM_006250376.1 | XP_006250438.1 | 5 | 1605 | |||
XM_006250377.2 | XP_006250439.1 | 4 | 1858 | |||
XM_008769824.1 | XP_008768046.1 | 2 | 1197 | |||
XM_008769825.1 | XP_008768047.1 | 2 | 3560 | |||
XM_008769826.1 | XP_008768048.1 | 4 | 2928 | |||
GenBank mRNA | FJ750588.1 | - | 1 | 366 |
