siRNA Details
Gene Symbol: Disp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105983.1 | NP_001099453.1 | 10 | 1854 | ||
XM_006250381.2 | XP_006250443.1 | 9 | 1723 | |||
XM_006250383.2 | XP_006250445.1 | 8 | 2184 | |||
XM_006250384.2 | XP_006250446.1 | 8 | 1663 | |||
XM_008769827.1 | XP_008768049.1 | 11 | 2548 | |||
XM_008769828.1 | XP_008768050.1 | 9 | 1834 |
