siRNA Details
Gene Symbol: Eprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024238.1 | NP_001019409.1 | 16, 17 | 1979 | ||
XM_006250425.2 | XP_006250487.1 | 17, 18 | 2232 | |||
XM_006250426.2 | XP_006250488.1 | 16, 17 | 2211 | |||
XM_006250427.2 | XP_006250489.1 | 13, 14 | 1612 | |||
GenBank mRNA | AY383705.1 | - | 16, 17 | 1979 |
