siRNA Details
Gene Symbol: Eprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024238.1 | NP_001019409.1 | 11 | 1180 | ||
XM_006250425.2 | XP_006250487.1 | 10 | 1316 | |||
XM_006250426.2 | XP_006250488.1 | 10 | 1316 | |||
XM_006250427.2 | XP_006250489.1 | 6 | 696 | |||
GenBank mRNA | AY383705.1 | - | 11 | 1180 |
