siRNA Details
Gene Symbol: Sdad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006958.1 | NP_001006959.1 | 18 | 1609 | ||
XM_006250722.2 | XP_006250784.1 | 18 | 1679 | |||
GenBank mRNA | BC083620.1 | - | 18 | 1609 | 7133993 |
