siRNA Details
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008295.1 | NP_001008296.1 | 8, 9 | 755 | ||
XM_006250884.2 | XP_006250946.1 | 8, 9 | 857 | |||
XM_006250887.2 | XP_006250949.1 | 8, 9 | 855 | |||
GenBank mRNA | BC085767.1 | - | 8, 9 | 755 | 7192362 | |
EU224276.1 | - | 8, 9 | 754 |
