siRNA Details
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106006.1 | NP_001099476.1 | 5 | 526 | ||
XM_006250934.2 | XP_006250996.1 | 5 | 723 | |||
XM_008770140.1 | XP_008768362.1 | 3 | 323 | |||
GenBank mRNA | FQ233155.1 | - | 5 | 474 | ||
FQ234076.1 | - | 5 | 504 |
