siRNA Details
Gene Symbol: LOC100910750 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751372.3 | XP_003751420.2 | 510 |
Gene Symbol: Dhx15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191597.1 | NP_001178526.1 | 4 | 1012 | ||
XM_006251031.2 | XP_006251093.1 | 4 | 1016 |
