siRNA Details
Gene Symbol: Lyar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011911.1 | NP_001011911.1 | 5 | 523 | ||
XM_006251087.1 | XP_006251149.1 | 5 | 529 | |||
GenBank mRNA | BC079008.1 | - | 5 | 523 | 7113133 |
