siRNA Details
Gene Symbol: Psmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198730.2 | NP_942025.1 | 1 | 70 | ||
GenBank mRNA | BC059159.1 | - | 1 | 75 | 6887177 | |
FQ212955.1 | - | 1 | 70 | |||
FQ213157.1 | - | 1 | 102 | |||
FQ213627.1 | - | 1 | 94 | |||
FQ214459.1 | - | 1 | 70 | |||
FQ215751.1 | - | 1 | 70 | |||
FQ229068.1 | - | 1 | 87 |
