siRNA Details
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011915.1 | NP_001011915.1 | 10 | 1305 | ||
XM_006251755.2 | XP_006251817.1 | 11 | 1363 | |||
XM_006251756.2 | XP_006251818.1 | 11 | 1317 | |||
XM_006251757.2 | XP_006251819.1 | 11 | 1363 | |||
XM_006251758.2 | XP_006251820.1 | 10 | 1330 | |||
XM_006251759.2 | XP_006251821.1 | 11 | 1363 | |||
XM_006251760.2 | XP_006251822.1 | 10 | 1330 | |||
XM_006251761.2 | XP_006251823.1 | 10 | 1327 | |||
GenBank mRNA | BC083876.1 | - | 10 | 1305 | 7129898 | |
FQ217336.1 | - | 9 | 1070 |
