siRNA Details
Gene Symbol: Cdkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106028.1 | NP_001099498.1 | 2 | 133 | ||
XM_006251791.2 | XP_006251853.1 | 2 | 164 | |||
XM_006251792.2 | XP_006251854.1 | 2 | 164 | |||
GenBank mRNA | BC167026.1 | - | 1 | 78 | 8375927 |
