siRNA Details
Gene Symbol: Efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106033.2 | NP_001099503.1 | 3 | 765 | ||
GenBank mRNA | BC161942.1 | - | 3 | 765 | 9023909 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106033.2 | NP_001099503.1 | 3 | 765 | ||
GenBank mRNA | BC161942.1 | - | 3 | 765 | 9023909 |