siRNA Details
Gene Symbol: Mphosph8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017375.2 | NP_001017375.2 | 12 | 2423 | ||
GenBank mRNA | BC099116.1 | - | 12 | 2366 | 7462184 | |
BP474651.1 | - | 3 | 462 |
