siRNA Details
Gene Symbol: LOC102551192 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773038.1 | XP_008771260.1 | 2 | 221 |
Gene Symbol: LOC102550885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006252233.2 | XP_006252295.1 | 3 | 312 |
Gene Symbol: LOC102551397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773039.1 | XP_008771261.1 | 221 |
