siRNA Details
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013051.1 | NP_001013069.1 | 3, 4 | 233 | ||
GenBank mRNA | BC158724.1 | - | 3, 4 | 233 | 9090831 | |
FQ220550.1 | - | 3, 4 | 268 | |||
FQ221379.1 | - | 3, 4 | 259 | |||
FQ225182.1 | - | 3, 4 | 351 | |||
FQ227975.1 | - | 3, 4 | 351 | |||
FQ232174.1 | - | 3, 4 | 351 |
