siRNA Details
Gene Symbol: Sugt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013051.1 | NP_001013069.1 | 5, 6 | 375 | ||
GenBank mRNA | AY318960.1 | - | 2, 3 | 120 | ||
BC158724.1 | - | 5, 6 | 375 | 9090831 | ||
FQ220550.1 | - | 5, 6 | 410 | |||
FQ221379.1 | - | 5, 6 | 401 | |||
FQ225182.1 | - | 5, 6 | 493 | |||
FQ227975.1 | - | 5, 6 | 493 | |||
FQ232174.1 | - | 5, 6 | 493 |
